missing translation for 'onlineSavingsMsg'
Learn More

L-Dihydroorotic acid, 99%

Product Code. 11906541 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11906541 1 g Glass bottle 1g
11916541 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11906541 Supplier Thermo Scientific Acros Supplier No. 437240010
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 5988-19-2 | C5H6N2O4 | 158.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5988-19-2
Molecular Formula C5H6N2O4
Molecular Weight (g/mol) 158.11
InChI Key UFIVEPVSAGBUSI-REOHCLBHSA-N
Synonym l-dihydroorotic acid, s-2,6-dioxohexahydropyrimidine-4-carboxylic acid, l-dihydroorotate, dihydro-l-orotic acid, s-dihydroorotic acid, l-hydroorotic acid, s-dihydroorotate, hydroorotic acid, l, 4s-2,6-dioxo-1,3-diazinane-4-carboxylic acid, s-4,5-dihydroorotate
PubChem CID 439216
ChEBI CHEBI:17025
IUPAC Name (4S)-2,6-dioxo-1,3-diazinane-4-carboxylic acid
SMILES C1C(NC(=O)NC1=O)C(=O)O

Specifications

Melting Point 254.0°C to 255.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 158.11
Percent Purity 99%
Physical Form Powder
Chemical Name or Material L-Dihydroorotic acid
compliance-icons
Product Identifier
  • L-Dihydroorotic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.