missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Dihydroorotic acid, 99%
Handla allt< titel> Produkter
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Packaging:
Glass bottle
Förpackningsstorlek
1g
5g
Beskrivning
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifikationer
Specifikationer
| Melting Point | 254.0°C to 255.0°C |
| Color | White |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98.5% min. (HPLC) |
| Packaging | Glass bottle |
| Quantity | 5 g |
| Formula Weight | 158.11 |
| Percent Purity | 99% |
| Physical Form | Powder |
| Chemical Name or Material | L-Dihydroorotic acid |
Säkerhet och hantering
Produktbeteckning
- L-Dihydroorotic acid
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering