missing translation for 'onlineSavingsMsg'
Learn More

L-DOPA, TRC

Product Code. 30371540
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30371540 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30371540 Supplier Toronto Research Chemicals Supplier No. TRCD5337515G
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Chemical Name or Material Levodopa
CAS 59-92-7
Purity Grade Notes HPLC
Molecular Formula C9 H11 N O4
Quantity 5 g
Synonym Levodopa, L-Tyrosine, 3-hydroxy- (9CI, ACI), 3-Hydroxy-L-tyrosine (ACI), Alanine, 3-(3,4-dihydroxyphenyl)-, L- (8CI), (-)-3,4-Dihydroxyphenylalanine, (-)-Dopa, (2S)-2-Amino-3-(3,4-dihydroxyphenyl)propanoic acid, (2S)-2-Amino-3-(3,4-dihydroxyphenyl)propanoic acid, (S)-3,4-Dihydroxyphenylalanine, 3,4-Dihydroxy-L-phenylalanine, 3,4-Dihydroxyphenyl-L-alanine, 3,4-Dihydroxyphenylalanine, 3-(3,4-Dihydroxyphenyl)-L-alanine, 3-Hydroxytyrosine, L-(-)-Dopa, L-3,4-Dihydroxyphenylalanine, L-3-(3,4-Dihydroxyphenyl)alanine, L-4,5-Dihydroxyphenylalanine, L-DOPA, L-β-(3,4-Dihydroxyphenyl)-α-alanine, Alphadopa, Bendopa, Brocadopa, Cidandopa, DA, Deadopa, Dihydroxy-L-phenylalanine, Dihydroxyphenylalanine, DOPA, Dopaflex, Dopaidan, Dopal, Dopalina, Dopar, Doparkine, Doparl, Dopasol, Dopaston, Dopaston SE, Dopastone, Dopastral, Dopicar, Doprin, Eldopal, Eldopar, Eldopatec, Eurodopa, Helfo-dopa, Inbrija, Insulamina, Laradopa, Larodopa, Ledopa, Levopa, Maipedopa, Parda, Pardopa, Syndopa, Veldopa, Weldopa, β-(3,4-Dihydroxyphenyl)-L-alanine, β-(3,4-Dihydroxyphenyl)-α-L-alanine, β-(3,4-Dihydroxyphenyl)alanine
InChI Formula InChI=1S/C9H11NO4/c10-6(9(13)14)3-5-1-2-7(11)8(12)4-5/h1-2,4,6,11-12H,3,10H2,(H,13,14)/t6-/m0/s1
SMILES N[C@@H](Cc1ccc(O)c(O)c1)C(=O)O
IUPAC Name (2S)-2-amino-3-(3,4-dihydroxyphenyl)propanoic acid
Molecular Weight (g/mol) 197.19
Formula Weight 197.0688
Percent Purity >95
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.