missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L(-)-Fucose, 97%

Product Code. 10523011 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 10523011

Brand: Thermo Scientific Chemicals undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2438-80-4 | C6H12O5 | 164.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2438-80-4
Molecular Formula C6H12O5
Molecular Weight (g/mol) 164.16
MDL Number MFCD00135607
InChI Key SHZGCJCMOBCMKK-DHVFOXMCSA-N
Synonym l---fucose, aldehydo-l-fucose, l-galactose, 6-deoxy, 2s,3r,4r,5s-2,3,4,5-tetrahydroxyhexanal, unii-28ryy2iv3f, l---rhodeose, 28ryy2iv3f, l--fucose, l-rhodeose, 3h-fucose
PubChem CID 3034656
ChEBI CHEBI:48204
SMILES C[C@@H]1OC(O)[C@@H](O)[C@H](O)[C@@H]1O

Specifications

Melting Point 128.0°C to 141.0°C
Color Beige to Gray-Purple
Loss on Drying 0.5% max. (60°C, 4 hrs) (vacuum)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 1 g
Beilstein 31, 78
Merck Index 15, 4308
Specific Rotation Condition - 78.00 (20.00°C c=4,H2O,NH3)
Specific Rotation − 78.00
Solubility Information Solubility in water: soluble in water
IUPAC Name (3S,4R,5S,6S)-6-methyloxane-2,3,4,5-tetrol
Formula Weight 164.16
Percent Purity 97%
Physical Form Powder
Chemical Name or Material L(-)-Fucose, 97%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.