missing translation for 'onlineSavingsMsg'
Learn More

L-Glucono-1,5-lactone, TRC

Product Code. 30240906
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30240906 500 mg 500mg
30240633 250 mg 250mg
30240878 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30240906 Supplier Toronto Research Chemicals Supplier No. TRCG413305500MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52153-09-0
Molecular Formula C6 H10 O6
Molecular Weight (g/mol) 178.14
Synonym L-Gluconic acid, δ-lactone, (3S,4R,5R,6S)-3,4,5-Trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-one, L-Glucono-1,5-lactone, L-Glucono-δ-lactone
IUPAC Name (3S,4R,5R,6S)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-one
SMILES OC[C@@H]1OC(=O)[C@@H](O)[C@H](O)[C@H]1O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C6H10O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-5,7-10H,1H2/t2-,3-,4+,5-/m0/s1
Formula Weight 178.0477
Chemical Name or Material L-Glucono-1,5-lactone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.