missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic acid 5-tert-butyl 1-methyl ester hydrochloride, 98%

Product Code. 15418436
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15418436

Brand: Thermo Scientific Alfa Aesar H63661.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6234-01-1 | C10H20ClNO4 | 253.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6234-01-1
Molecular Formula C10H20ClNO4
Molecular Weight (g/mol) 253.72
MDL Number MFCD00153438
InChI Key YIFPACFSZQWAQF-RZUXQYDTNA-N
Synonym h-glu otbu-ome.hcl, l-glutamic acid 5-tert-butyl 1-methyl ester hydrochloride, h-glu otbu-ome hcl, 5-tert-butyl 1-methyl l-glutamate hydrochloride, h-glu otbu-ome inverted exclamation mark currencyhcl, h-glu otbu-ome hydrochloride, s-5-tert-butyl 1-methyl 2-aminopentanedioate hydrochloride, h-glu otbu-omehcl, 5-tert-butyl 1-methyl 2s-2-aminopentanedioate hydrochloride, glu otbu-ome.hcl
PubChem CID 16218751
IUPAC Name 5-O-tert-butyl 1-O-methyl (2S)-2-aminopentanedioate;hydrochloride
SMILES CC(C)(C)OC(=O)CCC(C(=O)OC)N.Cl

Specifications

Beilstein 3572140
Quantity 1 g
Formula Weight 253.72
Percent Purity 98%
Physical Form Solid
Chemical Name or Material L-Glutamic acid 5-tert-butyl 1-methyl ester hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.