missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic acid, 98.5 to 101.5% (Dry Basis), Ultrapure

Product Code. 15865288 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
25 kg
Packaging:
Plastic Bottle
Unit Size:
1kg
25kg
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15865288 25 kg Plastic Bottle 25kg
15845288 1 kg Plastic Bottle 1kg
15855288 5 kg Plastic Bottle 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15865288 Supplier Thermo Scientific Supplier No. J16215.C4
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 56-86-0 | C5H9NO4 | 147.13 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-86-0
Molecular Formula C5H9NO4
Molecular Weight (g/mol) 147.13
MDL Number MFCD00002634
InChI Key WHUUTDBJXJRKMK-VKHMYHEASA-N
Synonym l-glutamic acid, glutamic acid, 2 s-2-aminopentanedioic acid, h-glu-oh, s-2-aminopentanedioic acid, glutaminol, l-glutaminic acid, glutacid, glutamicol, glutamidex
PubChem CID 33032
ChEBI CHEBI:16015
IUPAC Name (2 S)-2-aminopentanedioic acid
SMILES C(CC(=O)O)C(C(=O)O)N

Specifications

Melting Point 189°C to 192°C (decomposition)
Packaging Plastic Bottle
Linear Formula HOOCCH2CH2CH(NH2)COOH
Beilstein 1723801
Density 1.525
Quantity 25 kg
Formula Weight 147.13
Percent Purity 98.5 to 101.5%
Specific Gravity 1.525
Physical Form Crystalline Powder
Assay Dry basis
Chemical Name or Material L-Glutamic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.