missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic Acid, ≥99%, MP Biomedicals™

Product Code. 17837676 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
1 kg
5 kg
Unit Size:
0.1kg
1kg
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17837676 100 g 0.1kg
11374229 1 kg 1kg
11384229 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17837676 Supplier MP Biomedicals Supplier No. 02101793.1
  • ONLINE EXCLUSIVE PRICE
    32.10 €
    30.45 € / 0.1kg
    Save 1.65 €
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 04-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 56-86-0 | C5H9NO4 | 147.13 g/mol

Molecular biology, Cell Biology

Chemical Identifiers

CAS 56-86-0
Molecular Formula C5H9NO4
Molecular Weight (g/mol) 147.13
MDL Number MFCD00002634
InChI Key WHUUTDBJXJRKMK-UHFFFAOYNA-N
Synonym l-glutamic acid, glutamic acid, 2s-2-aminopentanedioic acid, h-glu-oh, s-2-aminopentanedioic acid, glutaminol, l-glutaminic acid, glutacid, glutamicol, glutamidex
PubChem CID 33032
ChEBI CHEBI:16015
SMILES NC(CCC(O)=O)C(O)=O

Specifications

Color White
Assay Percent Range ≥99%
Density 1.538g/mL at 20°C (Lit.)
IUPAC Name 2-aminopentanedioic acid
Quantity 100 g
Formula Weight 147.1
Grade Biochemical
Physical Form Powder
Chemical Name or Material L-Glutamic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.