missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic acid diethyl ester hydrochloride, 98%, Thermo Scientific™

Product Code. 15452262 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15452262 5 g 5g
15482262 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15452262 Supplier Thermo Scientific Acros Supplier No. 459260050
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 1118-89-4
Molecular Formula C9H17NO4·HCl
Molecular Weight (g/mol) 239.7
InChI Key WSEQLMQNPBNMSL-FJXQXJEOSA-N
Synonym l-glutamic acid diethyl ester hydrochloride, diethyl l-glutamate hydrochloride, h-glu oet-oet.hcl, diethyl glutamate hydrochloride, h-glu oet-oet hcl, s-diethyl 2-aminopentanedioate hydrochloride, diethyl l-glutaminate hydrochloride, 1,5-diethyl 2s-2-aminopentanedioate hydrochloride, l-glutamic acid, diethyl ester, hydrochloride, diethyl 2s-2-aminopentanedioate hydrochloride
PubChem CID 73960
IUPAC Name diethyl (2S)-2-aminopentanedioate;hydrochloride
SMILES CCOC(=O)CCC(C(=O)OCC)N.Cl

Specifications

Melting Point 108.0°C to 110.0°C
Infrared Spectrum Authentic
Quantity 5 g
Solubility Information (5% in water) Clear colorless
Formula Weight 239.7
Percent Purity 98%
Chemical Name or Material L-Glutamic acid diethyl ester hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.