missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic acid dimethyl ester hydrochloride, 98+%

Product Code. 11356646
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11356646 25 g 25g
11346646 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11356646 Supplier Thermo Scientific Alfa Aesar Supplier No. L03764.14
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 23150-65-4 | C7H13ClNO4 | 210.63 g/mol

L-Glutamic acid dimethyl ester hydrochloride is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Glutamic acid dimethyl ester hydrochloride is used as a pharmaceutical intermediate.

Solubility
Soluble in water, methanol (50 mg/ml).

Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, water/ moisture, heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23150-65-4
Molecular Formula C7H13ClNO4
Molecular Weight (g/mol) 210.63
MDL Number MFCD00038879
InChI Key BPHCSYSXOKTCMA-UHFFFAOYNA-N
Synonym l-glutamic acid dimethyl ester hydrochloride, h-glu ome-ome.hcl, dimethyl l-glutamate hydrochloride, s-dimethyl 2-aminopentanedioate hydrochloride, unii-185vc5x3pf, 1,5-dimethyl 2s-2-aminopentanedioate hydrochloride, h-glu ome-ome inverted exclamation mark currencyhcl, s-dimethyl 2-aminopentanedioate hcl, glutamic acid dimethyl ester hydrochloride, dimethyl glutamic acid hydrochloride
PubChem CID 12917568
IUPAC Name dimethyl (2S)-2-aminopentanedioate;hydrochloride
SMILES [Cl].COC(=O)CCC(N)C(=O)OC

Specifications

Melting Point 99°C to 103°C
Beilstein 4238829
Solubility Information Soluble in water,methanol (50mg/ml).
Quantity 25 g
Odor Odorless
Formula Weight 211.65
Optical Rotation +26° (c=5 in Water)
Sensitivity Hygroscopic
Percent Purity ≥98%
Chemical Name or Material L-Glutamic acid dimethyl ester hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.