missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic Acid Dimethyl Ester Hydrochloride, TRC

Product Code. 30392853
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30392853 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30392853 Supplier Toronto Research Chemicals Supplier No. TRCG59697010G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dimethyl L-Glutamate Hydrochloride
CAS 23150-65-4
Molecular Formula C7 H13 N O4 . Cl H
Quantity 10 g
Synonym L-Glutamic acid, 1,5-dimethyl ester, hydrochloride (1:1), Glutamic acid, dimethyl ester, hydrochloride, L- (7CI,8CI), L-Glutamic acid, dimethyl ester, hydrochloride (9CI), Dimethyl (S)-2-aminopentanedioate hydrochloride, Dimethyl L-glutamate hydrochloride, Dimethyl glutamate hydrochloride, Glutamic acid dimethyl ester hydrochloride, NSC 83254
InChI Formula InChI=1S/C7H13NO4.ClH/c1-11-6(9)4-3-5(8)7(10)12-2;/h5H,3-4,8H2,1-2H3;1H/t5-;/m0./s1
SMILES Cl.COC(=O)CC[C@H](N)C(=O)OC
IUPAC Name dimethyl (2S)-2-aminopentanedioate;hydrochloride
Molecular Weight (g/mol) 211.6434
Formula Weight 211.0611
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.