missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Glutamic acid hydrochloride, 99%

Product Code. 11424154
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11424154 250 g 250g
11414154 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11424154 Supplier Thermo Scientific Chemicals Supplier No. A12505.30
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 138-15-8 | C5H10ClNO4 | 183.588 g/mol

L-Glutamic acid hydrochloride is used as a digestive aid. It is used in cell culture as a component of MEM non-essential amino acids solution. It has been used as a nitrogen source in the culture of Aspergillus fumigatus NRRL 2436 for fumagillin production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Glutamic acid hydrochloride is used as a digestive aid. It is used in cell culture as a component of MEM non-essential amino acids solution. It has been used as a nitrogen source in the culture of Aspergillus fumigatus NRRL 2436 for fumagillin production.

Solubility
Soluble in water (490 g/L).

Notes
Hygroscopic. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138-15-8
Molecular Formula C5H10ClNO4
Molecular Weight (g/mol) 183.588
MDL Number MFCD00012619
InChI Key RPAJSBKBKSSMLJ-DFWYDOINSA-N
Synonym l-glutamic acid hydrochloride, s-2-aminopentanedioic acid hydrochloride, l-+-glutamic acid hydrochloride, glutamic acid hydrochloride, unii-m0c2sp444t, glu hcl, acidogen, acidulin, gastuloric, glutamidin
PubChem CID 2723891
IUPAC Name (2S)-2-aminopentanedioic acid;hydrochloride
SMILES C(CC(=O)O)C(C(=O)O)N.Cl

Specifications

Melting Point 203°C (decomposition)
Beilstein 3565569
Merck Index 14,4469
Solubility Information Soluble in water (490g/L).
Density 1.525
Quantity 250 g
Formula Weight 183.59
Optical Rotation +23° (c=6 in Water)
Sensitivity Hygroscopic
Percent Purity 99%
Chemical Name or Material L-Glutamic acid hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • L-Glutamic acid hydrochloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: L-Glutamic acid, hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.