missing translation for 'onlineSavingsMsg'
Learn More

l-Glutamic Acid, Millipore™

Product Code. 10742062
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10742062 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10742062 Supplier Millipore Supplier No. 1002910250
 Restricted Item
Estimated Shipment: 16-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For biochemistry

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-86-0

Specifications

Melting Point 160°C
MDL Number MFCD00002634
Synonym L-Glutamic acid, L-α-Aminoglutaric acid, Glu, L-alpha-Aminoglutaric acid, L-Glutamic acid
Density 1.54 g/cm3 at 20°C
Molecular Weight (g/mol) 147.13
Quantity 250 g
Grade Research
Chemical Name or Material L-Glutamic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.