missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic acid monopotassium salt monohydrate, 97+%

Product Code. 11413457
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11413457 1000 g 1000g
11423457 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11413457 Supplier Thermo Scientific Alfa Aesar Supplier No. A17232.0B
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6382-01-0 | C5H11KNO5 | 204.243 g/mol

L-Glutamic acid monopotassium salt monohydrate acts as a common excitatory neurotransmitter like the agonist at kainate, N-methyl-D-aspartate receptor (NMDA) and quisqualate receptors in central nervous system. It is also used in cell culture as a component of MEM non-essential amino acids solution..

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Glutamic acid monopotassium salt monohydrate acts as a common excitatory neurotransmitter like the agonist at kainate, N-methyl-D-aspartate receptor (NMDA) and quisqualate receptors in central nervous system. It is also used in cell culture as a component of MEM non-essential amino acids solution..

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6382-01-0
Molecular Formula C5H11KNO5
Molecular Weight (g/mol) 204.243
MDL Number MFCD00150343
InChI Key VMWJTBYKMYBHOC-QTNFYWBSSA-N
Synonym l-glutamic acid hydrate potassium, l-glutamicacid,monopotassiumsalt,monohydrate
PubChem CID 87237506
IUPAC Name (2S)-2-aminopentanedioic acid;potassium;hydrate
SMILES C(CC(=O)O)C(C(=O)O)N.O.[K]

Specifications

Melting Point 115°C to 118°C
Solubility Information Soluble in water.
Quantity 1000 g
Formula Weight 203.25 (185.23 Anhydrous)
Percent Purity ≥97%
Chemical Name or Material L-Glutamic acid monopotassium salt monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.