missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L(+)-Glutamic acid monosodium salt monohydrate, 99%

Product Code. 10265390 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
Plastic Bottle
Plastic Drum
Unit Size:
10kg
1kg
2.5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10265390 1 kg Plastic Bottle 1kg
10142770 2.5 kg Plastic Bottle 2.5kg
10237543 10 kg Plastic Drum 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10265390 Supplier Thermo Scientific Chemicals Supplier No. 119940010
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6106-04-03 | C5H8NNaO4·H2O | 187.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6106-04-03
Molecular Formula C5H8NNaO4·H2O
Molecular Weight (g/mol) 187.13
MDL Number MFCD00150138
InChI Key GJBHGUUFMNITCI-QTNFYWBSSA-M

Specifications

Melting Point 232.0°C
Color White
Chloride 2000ppm max
Infrared Spectrum Authentic
Assay Percent Range 0.99
Loss on Drying 0.5% max. (100°C, 5 hrs)
Packaging Plastic Bottle
Linear Formula NaO2CCH2CH2CH(NH2)CO2H·H2O
Specific Rotation + 25.00
Sulfate 500ppm max
Synonym l-thyroxine sodium salt pentahydrate, l-thyroxine sodium pentahydrate, sodium l-thyroxine pentahydrate, sodium levothyroxine, levothyroxine sodium, 3-4-4-hydroxy-3,5-diiodophenoxy-3,5-diiodophenyl-l-alanine sodium salt, levothyroxine sodium pentahydrate, l-tyrosine, o-4-hydroxy-3,5-diiodophenyl-3,5-diiodo-, sodium salt, pentahydrate, o-4-hydroxy-3,5-diiodophenyl-3,5-diiodo-l-tyrosine sodium salt pentahydrate
Solubility Information Solubility in water: soluble. Other solubilities: sparingly soluble in alcohol
SMILES O.[Na+].N[C@@H](CCC([O-])=O)C(O)=O
IUPAC Name sodium (4S)-4-amino-4-carboxybutanoate hydrate
Specific Rotation Condition + 25.00 (20.00°C c=10,2 N HCl)
Quantity 1 kg
PubChem CID 23665037
Formula Weight 187.13
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material L(+)-Glutamic acid monosodium salt monohydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.