missing translation for 'onlineSavingsMsg'
Learn More

L-(+)-Glutamic Acid, Monosodium Salt, Monohydrate, BAKER™, J.T. Baker™

Product Code. 15508794 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15508794 500 g Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15508794 Supplier Avantor J.T.Baker Supplier No. M74607
  • ONLINE EXCLUSIVE PRICE
    134.00 €
    79.24 € / Pack of 1
    Save 54.76 €
    40% Off
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A grade of chemicals for laboratory use

Specifications

CAS 6106-04-3
Molecular Formula C5H10NNaO5
Packaging Poly Bottle
Synonym monosodium glutamate, l-glutamic acid monosodium salt monohydrate, p.a, l-glutamic acid monosodium salt monohydrate nt, l-glutamic acid monosodium salt monohydrate, saj special grade, monosodium glutamate, united states pharmacopeia usp reference standard, l-glutamic acid, monosodium salt, low endotoxin, pharmagrade, manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production., suitable for cell culture
InChI Key GJBHGUUFMNITCI-QTNFYWBSSA-M
SMILES C(CC(=O)O)C(C(=O)O)N.O.[Na]
IUPAC Name (2S)-2-aminopentanedioic acid;sodium;hydrate
Molecular Weight (g/mol) 187.13
Quantity 500 g
PubChem CID 87090819
Formula Weight 187.13
Percent Purity 98.5 to 101.5%
Grade FCC
Chemical Name or Material L-(+)-Glutamic Acid, Monosodium Salt, Monohydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.