missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic Acid, Multi-Compendial, F.C.C., J.T. Baker™

Product Code. 15507964 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15507964 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15507964 Supplier Avantor J.T.Baker Supplier No. 207706
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity, multi-compendial L-Glutamic acid suitable for manufacturing, processing, or repackaging applications.

Specifications

CAS 56-86-0
Molecular Formula C5H9NO4
Packaging Poly Bottle
MDL Number MFCD00002634
Synonym l-glutamic acid, glutamic acid, 2s-2-aminopentanedioic acid, h-glu-oh, s-2-aminopentanedioic acid, glutaminol, l-glutaminic acid, glutacid, glutamicol, glutamidex
InChI Key WHUUTDBJXJRKMK-UHFFFAOYNA-N
SMILES NC(CCC(O)=O)C(O)=O
IUPAC Name (2S)-2-aminopentanedioic acid
Molecular Weight (g/mol) 147.13
Quantity 1 kg
PubChem CID 33032
ChEBI CHEBI:16015
Formula Weight 147.13
Percent Purity 98.5 to 101.5%
Specific Gravity 1L = 1.54kg
Grade FCC/Multi-Compendial
Chemical Name or Material L-Glutamic Acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.