missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamine, ≥98.5%, Ultrapure

Product Code. 15875148 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
Packaging:
Plastic Bottle
Unit Size:
100g
1kg
250g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15875148 100 g Plastic Bottle 100g
15885148 250 g Plastic Bottle 250g
15895148 500 g Plastic Bottle 500g
15875288 1 kg Plastic Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15875148 Supplier Thermo Scientific Supplier No. J16285.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 56-85-9 | C5H10N2O3 | 146.146 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-85-9
Molecular Formula C5H10N2O3
Molecular Weight (g/mol) 146.146
MDL Number MFCD00008044
InChI Key ZDXPYRJPNDTMRX-VKHMYHEASA-N
Synonym l-glutamine, glutamine, levoglutamide, l-+-glutamine, glutamic acid amide, h-gln-oh, stimulina, cebrogen, glumin, levoglutamid
PubChem CID 5961
ChEBI CHEBI:18050
IUPAC Name (2 S)-2,5-diamino-5-oxopentanoic acid
SMILES C(CC(=O)N)C(C(=O)O)N

Specifications

Packaging Plastic Bottle
Linear Formula H2NCOCH2CH2CH(NH2)COOH
Beilstein 1723797
Quantity 100 g
Formula Weight 146.14
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Chemical Name or Material L-Glutamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.