missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamine, 99%

Product Code. 11492277
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11492277 250 g 250g
11482277 50 g 50g
11472277 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11492277 Supplier Thermo Scientific Alfa Aesar Supplier No. A14201.30
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Involved in many metabolic processes and regulates acid-base balance in the kidney. It is an important amino acid for the incorporation of ammonium ions into biomolecules. It is a vital component of culture media that serves as a major energy source for cells in culture. | CAS: 56-85-9 | C5H10N2O3 | 146.15 g/mol

L-Glutamine is involved in many metabolic processes and regulates acid-base balance in the kidney. It is an important amino acid for the incorporation of ammonium ions into biomolecules. It is a vital component of culture media that serves as a major energy source for cells in culture. As a component in oral supplementation, it is useful for patients who have advanced stages of cancer to reverse cachexia (i.e. muscle wasting). It is also used for attention deficit-hyperactivity disorder (ADHD), a urinary condition called cystinuria, sickle cell anemia and for alcohol withdrawal support. It finds applications in patients with HIV (AIDS) to prevent weight loss.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Glutamine is involved in many metabolic processes and regulates acid-base balance in the kidney. It is an important amino acid for the incorporation of ammonium ions into biomolecules. It is a vital component of culture media that serves as a major energy source for cells in culture. As a component in oral supplementation, it is useful for patients who have advanced stages of cancer to reverse cachexia (i.e. muscle wasting). It is also used for attention deficit-hyperactivity disorder (ADHD), a urinary condition called cystinuria, sickle cell anemia and for alcohol withdrawal support. It finds applications in patients with HIV (AIDS) to prevent weight loss.

Solubility
Soluble in water, dimethyl sulfoxide and ethanol. Insoluble in methanol, ether, benzene, acetone, ethyl acetate and chloroform.

Notes
Moisture and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with moisture and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-85-9
Molecular Formula C5H10N2O3
Molecular Weight (g/mol) 146.15
MDL Number MFCD00008044
InChI Key ZDXPYRJPNDTMRX-VKHMYHEASA-N
Synonym l-glutamine, glutamine, levoglutamide, l-+-glutamine, glutamic acid amide, h-gln-oh, stimulina, cebrogen, glumin, levoglutamid
PubChem CID 5961
ChEBI CHEBI:18050
SMILES N[C@@H](CCC(N)=O)C(O)=O

Specifications

Melting Point 185°C (decomposition)
Packaging Plastic Bottle
Beilstein 1723797
Solubility Information Soluble in water,dimethyl sulfoxide and ethanol. Insoluble in methanol,ether,benzene,acetone,ethyl acetate and chloroform.
IUPAC Name (2S)-2-amino-4-carbamoylbutanoic acid
Quantity 250 g
Formula Weight 146.15
Percent Purity 99%
Physical Form Powder
Chemical Name or Material L-Glutamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.