missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamine, Extra Pure, SLR, Fisher Chemical

Product Code. 10030150 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10030150 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10030150 Supplier Fisher Chemical Supplier No. G/0514/50
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point 185°C
CAS 56-85-9
pH 4.0 to 6.0
Color White
Molecular Formula C5H10N2O3
Packaging Plastic Powder Jar
MDL Number MFCD00008044
Synonym l-glutamine, glutamine, levoglutamide, l-+-glutamine, glutamic acid amide, h-gln-oh, stimulina, cebrogen, glumin, levoglutamid
InChI Key ZDXPYRJPNDTMRX-VKHMYHEASA-N
SMILES N[C@@H](CCC(N)=O)C(O)=O
IUPAC Name (2S)-2,5-diamino-5-oxopentanoic acid
Molecular Weight (g/mol) 146.15
Quantity 250 g
PubChem CID 5961
ChEBI CHEBI:18050
Formula Weight 146.15g/mol
Physical Form Solid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.