missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamine, MP Biomedicals™

Product Code. 17807496 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17807496 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17807496 Supplier MP Biomedicals Supplier No. 0210180625
  • ONLINE EXCLUSIVE PRICE
    39.85 €
    37.80 € / 25g
    Save 2.05 €
    5% Off
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Participates in acid-base regulation in vivo. L-Glutamine, MP Biomedicals is a crucial component of culture media and serves as a major energy source for cells in culture. It is the uncharged, amidated analog of L-glutamic acid, an amino acid important fo

L-Glutamine is very stable as a dry powder and as a frozen solution. In liquid media or stock solutions, however, L-glutamine degrades relatively rapidly. Optimal cell performance usually requires supplementation of the media with L-glutamine prior to use.

  • L-glutamine is added for optimal cell performance in a culture
  • The utilization of glutamine under carbon starvation conditions by cultured Saccharomyces cerevisiae and its relation to the cytoplasmic compartmentation of the transcriptional activator Gln3 has been studied
  • The use of glutamine in cultured rat islets to probe a-ketoisocaproate oxidation has been investigated

Specifications

CAS 56-85-9
Molecular Formula C5H10N2O3
MDL Number MFCD00008044
Synonym l-glutamine, glutamine, levoglutamide, l-+-glutamine, glutamic acid amide, h-gln-oh, stimulina, cebrogen, glumin, levoglutamid
Solubility Information Soluble in water.
InChI Key ZDXPYRJPNDTMRX-VKHMYHEASA-N
SMILES N[C@@H](CCC(N)=O)C(O)=O
IUPAC Name (2S)-2-amino-4-carbamoylbutanoic acid
Molecular Weight (g/mol) 146.15
Quantity 25 g
PubChem CID 5961
ChEBI CHEBI:18050
Formula Weight 146.1
Percent Purity ≥99%
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.