missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamine, MP Biomedicals

Product Code. 11394249 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11394249 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11394249 Supplier MP Biomedicals Supplier No. 0210180691
  • ONLINE EXCLUSIVE PRICE
    604.00 €
    572.25 € / 1kg
    Save 31.75 €
    5% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity powder for use in research applications.

  • L-glutamine is an essential amino acid, which serves as a major energy source for cells in culture
  • L-glutamine is added for optimal cell performance in a culture medium
  • The utilization of glutamine under carbon starvation conditions by cultured Saccharomyces cerevisiae and its relation to the cytoplasmic compartmentation of the transcriptional activator Gln3 has been studied
  • The use of glutamine in cultured rat islets to probe a-ketoisocaproate oxidation has been investigated

Specifications

Melting Point 185°C (literature)
CAS 56-85-9
Color White
Molecular Formula C5H10N2O3
Linear Formula C5H10N2O3
MDL Number MFCD00008044
Synonym l-glutamine, glutamine, levoglutamide, l-+-glutamine, glutamic acid amide, h-gln-oh, stimulina, cebrogen, glumin, levoglutamid
Solubility Information Soluble in water.
InChI Key ZDXPYRJPNDTMRX-VKHMYHEASA-N
SMILES N[C@@H](CCC(N)=O)C(O)=O
IUPAC Name (2S)-2,5-diamino-5-oxopentanoic acid
Molecular Weight (g/mol) 146.15
Quantity 1 kg
PubChem CID 5961
ChEBI CHEBI:18050
Formula Weight 146.1
Percent Purity ≥99%
Physical Form Crystalline Powder
Chemical Name or Material L-GLUTAMINE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.