missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamine, MP Biomedicals

Product Code. 11464795 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11464795 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11464795 Supplier MP Biomedicals Supplier No. 091680149
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

200 mM solution Shipped frozen in dry ice

Specifications

CAS 56-85-9
Molecular Formula C5H10N2O3
MDL Number MFCD00008044
Synonym l-glutamine, glutamine, levoglutamide, l-+-glutamine, glutamic acid amide, h-gln-oh, stimulina, cebrogen, glumin, levoglutamid
InChI Key ZDXPYRJPNDTMRX-VKHMYHEASA-N
SMILES N[C@@H](CCC(N)=O)C(O)=O
IUPAC Name (2S)-2-amino-4-carbamoylbutanoic acid
Molecular Weight (g/mol) 146.15
Quantity 100 mL
PubChem CID 5961
ChEBI CHEBI:18050
Grade Cell Culture
Chemical Name or Material L-Glutamine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.