missing translation for 'onlineSavingsMsg'
Learn More

L(+)-Glutamine, specified according to the requirements of USP

Product Code. 15229295 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15229295 50 g 50g
15239295 250 g 250g
15219295 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15229295 Supplier Thermo Scientific Acros Supplier No. 386030500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 56-85-9 | C5H10N2O3 | 146.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-85-9
Molecular Formula C5H10N2O3
Molecular Weight (g/mol) 146.15
MDL Number MFCD00008044
InChI Key ZDXPYRJPNDTMRX-VKHMYHEASA-N
Synonym l-glutamine, glutamine, levoglutamide, l-+-glutamine, glutamic acid amide, h-gln-oh, stimulina, cebrogen, glumin, levoglutamid
PubChem CID 5961
ChEBI CHEBI:18050
IUPAC Name (2S)-2,5-diamino-5-oxopentanoic acid
SMILES N[C@@H](CCC(N)=O)C(O)=O

Specifications

Melting Point 185.0°C
Linear Formula H2NCOCH2CH2CH(NH2)CO2H
Solubility Information Solubility in water: 35g/L (20°C). Other solubilities: practically insoluble in methanol, ethanol, ether,, benzene, acetone, ethyl acetate, chloroform, soluble in mineral acids, soluble in ethanol: 46 μg/100mL (25°C), soluble in methanol: 3.5 mg/100mL (2
Quantity 50 g
Formula Weight 146.15
Chemical Name or Material L(+)-Glutamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.