missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamine, TRC

Product Code. 30005310
Change view
Click to view available options
Quantity:
10 mg
50 mg
1 g
Unit Size:
10mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30005310 50 mg 50mg
30003890 1 g 1g
17947904 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30005310 Supplier Toronto Research Chemicals Supplier No. TRCG59700050MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-85-9
Molecular Formula C5 H10 N2 O3
Molecular Weight (g/mol) 146.14
Synonym L-Glutamine, (2S)-2,5-Diamino-5-oxopentanoic Acid, Aspartic Acid Imp. F (Pharmeuropa), 2-Aminoglutaramic acid, Acustasin, Aesgen 14, Cebrogen, Glumin, Glumin (amino acid), Glutamic acid 5-amide, Glutamic acid amide, Glutamine, L-(+)-Glutamine, L-2-Aminoglutaramidic acid, L-Glutamic acid γ-amide, Levoglutamide, NSC 27421, (S)-2,5-Diamino-5-oxopentanoic acid, Saforis, Stimulina, γ-Glutamine
IUPAC Name (2S)-2,5-diamino-5-oxopentanoic acid
SMILES N[C@@H](CCC(=O)N)C(=O)O

Specifications

InChI Formula InChI=1S/C5H10N2O3/c6-3(5(9)10)1-2-4(7)8/h3H,1-2,6H2,(H2,7,8)(H,9,10)/t3-/m0/s1
Quantity 50 mg
Formula Weight 146.0691
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material L-Glutamine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.