missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamine, USP Grade, MP Biomedicals™

Product Code. 11408382 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11408382 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11408382 Supplier MP Biomedicals Supplier No. 0219960890
 Restricted Item
Estimated Shipment: 25-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 56-85-9 | C5H10N2O3 | 146.15 g/mol

Chemical Identifiers

CAS 56-85-9
Molecular Formula C5H10N2O3
Molecular Weight (g/mol) 146.15
MDL Number MFCD00008044
InChI Key ZDXPYRJPNDTMRX-VKHMYHEASA-N
Synonym (S)-2,5-Diamino-5-oxopentanoic acid; L-Glutamic acid 5-amide
IUPAC Name (2S)-2-amino-4-carbamoylbutanoic acid
SMILES N[C@@H](CCC(N)=O)C(O)=O

Specifications

Melting Point 185.5°C
pH 5 to 6 (at 14.6 g/L, 25°C)
Color White
Beilstein 1723797
Density 1.364 g/cm3
Quantity 500 g
Optical Rotation (40 mg/mL aq. soln. (at 20°C) +6.3° to +7.3°)
Grade USP
Physical Form Powder
Chemical Name or Material L-Glutamine, USP

For research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.