missing translation for 'onlineSavingsMsg'
Learn More

γ-L-Glutamyl-p-nitroanilide, Hydrochloride, Fisher BioReagents

Product Code. 10450565
Change view
Click to view available options
Quantity:
500 mg
5 g
25 g
Packaging:
Amber Glass
Unit Size:
25g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10450565 500 mg Amber Glass 500mg
10596374 5 g Amber Glass 5g
10673285 25 g Amber Glass 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10450565 Supplier Fisher BioReagents Supplier No. BP2656500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
γ-L-Glutamyl-p-Nitroanilide, Hydrochloride is a water soluble form of GPNA suitable for use as substrate for γ-glutamyl transpeptidase.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67953-08-6
Molecular Formula C11H14ClN3O5
Molecular Weight (g/mol) 303.70
MDL Number MFCD00039054
InChI Key WMRCHNNHOJNRSL-UHFFFAOYNA-N
Synonym gamma-l-glutamyl-p-nitroanilide, n-4-nitrophenyl-l-glutamine monohydrochloride, l-2-amino-4'-nitroglutaranilic acid hydrochloride, 2-amino-4-4-nitrophenyl carbamoyl butanoic acid hydrochloride, l-, a-glutamyl-p-nitroanilide, l-glutamic acid, a-p-nitroanilide hydrochloride, l-glutamic acid gamma-p-nitroanilide hydrochlori, l-glutamic acid gamma-p-nitroanilide hydrochloride, gamma-glutamyl transpeptidase substrate
IUPAC Name N-(3-carbamoyl-1-carboxypropyl)-4-nitroanilinium chloride
SMILES [Cl-].NC(=O)CCC([NH2+]C1=CC=C(C=C1)[N+]([O-])=O)C(O)=O

Specifications

Color Beige
Packaging Amber Glass
Quantity 500 mg
Formula Weight 303.7
Physical Form Powder or Solid
Chemical Name or Material γ-L-Glutamyl-p-Nitroanilide, Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.