missing translation for 'onlineSavingsMsg'
Learn More

L-Glutathione, reduced, 97%

Product Code. 11479146
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11479146 25 g 25g
11469146 5 g 5g
11489146 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11479146 Supplier Thermo Scientific Alfa Aesar Supplier No. A18014.14
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 70-18-8 | C10H17N3O6S | 307.321 g/mol

L-Glutathione is used in the treatment of lung diseases for patients who are HIV positive. It protects the cancerous cells by conferring resistance to chemotherapeutic drugs. It is involved in many aspects of metabolism including transport of g-glutanyl amino acids and reductive cleavage of disulfide bonds. As an antioxidant, it prevents damage to important cellular components that arise due to reactive oxygen species like free radicals and peroxide. It is also used to decrease the concentrations of inflammatory cytokines (IL-6, IL-18) as well as involved in increasing the level of serum Ca2+ ions. It is also used in white wine production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water, dimethyl formamide, ethanol, phosphate buffered saline, diluted alcohol, liquid ammonia and dimethyl sulfoxide.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70-18-8
Molecular Formula C10H17N3O6S
Molecular Weight (g/mol) 307.321
MDL Number MFCD00065939
InChI Key RWSXRVCMGQZWBV-WDSKDSINSA-N
Synonym glutathione, l-glutathione, glutathion, glutathione-sh, glutinal, isethion, tathion, reduced glutathione, deltathione, neuthion
PubChem CID 124886
ChEBI CHEBI:16856
IUPAC Name (2S)-2-amino-5-[[(2R)-1-(carboxymethylamino)-1-oxo-3-sulfanylpropan-2-yl]amino]-5-oxopentanoic acid
SMILES C(CC(=O)NC(CS)C(=O)NCC(=O)O)C(C(=O)O)N

Specifications

Melting Point ∽193°C (decomposition)
Assay Percent Range 0.97
Quantity 25 g
Beilstein 1729812
Merck Index 14,4475
Solubility Information Soluble in water,dimethyl formamide,ethanol,phosphate buffered saline,diluted alcohol,liquid ammonia and dimethyl sulfoxide.
Formula Weight 307.33
Percent Purity 97%
Chemical Name or Material L-Glutathione, Reduced

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.