missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Gulono-1,4-lactone, MedChemExpress
Description
L-Gulono-1,4-lactone is a substrate of L-gulono-1,4-lactone oxidoreductase, which catalyzes the last step of the biosynthesis of L-ascorbic (Vatamin) C. In other words, L-Gulono-1,4-lactone is a direct precursor of vitamin C in animals, in plants and in some protists.
Specifications
Specifications
| Chemical Name or Material | L-Gulono-1,4-lactone |
| CAS | 1128-23-0 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C6H10O6 |
| Quantity | 1 g |
| Solubility Information | H2O : 50 mg/mL (280.68 mM; Need ultrasonic) |
| SMILES | O=C1O[C@H]([C@@H](O)CO)[C@H](O)[C@@H]1O |
| Molecular Weight (g/mol) | 178.14 |
| Formula Weight | 178.14 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
¿Detecta una oportunidad de mejora?