missing translation for 'onlineSavingsMsg'
Learn More

L-Gulono-1,4-lactone, TRC

Product Code. 30136670
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136670 100 g 100g
30136344 25 g 25g
30007442 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136670 Supplier Toronto Research Chemicals Supplier No. TRCG855000100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1128-23-0
Molecular Formula C6 H10 O6
Molecular Weight (g/mol) 178.14
Synonym L-Gulonic acid, γ-lactone, Gulonic acid, γ-lactone, L- (8CI), Dihydroascorbic acid, L-Gulono-1,4-lactone, L-Gulono-γ-lactone, L-Gulonolactone, Reduced ascorbate, Reduced ascorbic acid, γ-Gulonolactone
IUPAC Name (3S,4R,5R)-5-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxyoxolan-2-one
SMILES OC[C@H](O)[C@H]1OC(=O)[C@@H](O)[C@H]1O

Specifications

Quantity 100 g
InChI Formula InChI=1S/C6H10O6/c7-1-2(8)5-3(9)4(10)6(11)12-5/h2-5,7-10H,1H2/t2-,3+,4-,5+/m0/s1
Formula Weight 178.0477
Percent Purity >95%
Chemical Name or Material L-Gulono-1,4-lactone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.