missing translation for 'onlineSavingsMsg'
Learn More

L-Histidine, Cell Culture Reagent, Thermo Scientific™

Product Code. 15405339
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
1 kg
Unit Size:
100g
10g
1kg
500g
Product Code. Quantity unitSize
15405339 10 g 10g
15415339 100 g 100g
15425339 500 g 500g
15435339 1 kg 1kg
4 options
This item is not returnable. View return policy

Product Code. 15405339

Brand: Thermo Scientific Alfa Aesar J63065.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-00-1
Molecular Formula C6H9N3O2
Molecular Weight (g/mol) 155.16
MDL Number MFCD00064315
InChI Key HNDVDQJCIGZPNO-UHFFFAOYNA-N
Synonym l-histidine, histidine, h-his-oh, glyoxaline-5-alanine, anti-rheuma, l---histidine, istidina, s-histidine, l-histidin
PubChem CID 6274
ChEBI CHEBI:15971
SMILES NC(CC1=CN=CN1)C(O)=O

Specifications

Melting Point ∽281°C (decomposition)
pH 7.0 to 8
Color White
Beilstein 84088
Merck Index 14,4720
IUPAC Name (2S)-2-amino-3-(1H-imidazol-5-yl)propanoic acid
Quantity 10 g
Odor Odorless
Formula Weight 155.16
Optical Rotation −39° (c=5 in Water)
Grade Cell Culture Reagent
Physical Form Powder
Chemical Name or Material L-Histidine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.