missing translation for 'onlineSavingsMsg'
Learn More

L-Histidine hydrochloride monohydrate, 98%

Product Code. 10761442 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic Bottle
Unit Size:
100g
2.5kg
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10761442 25 g Plastic Bottle 25g
10095790 100 g Plastic Bottle 100g
10444045 500 g Plastic Bottle 500g
10277563 2.5 kg Plastic Bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10761442 Supplier Thermo Scientific Acros Supplier No. 411730250
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 5934-29-2 | C6H12ClN3O3 | 209.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5934-29-2
Molecular Formula C6H12ClN3O3
Molecular Weight (g/mol) 209.63
MDL Number MFCD00151027
InChI Key CMXXUDSWGMGYLZ-XRIGFGBMSA-N
Synonym l-histidine hydrochloride hydrate, l-histidine hydrochloride monohydrate, s-2-amino-3-1h-imidazol-4-yl propanoic acid hydrochloride hydrate, h-his-oh.hcl.h2o, l-histidine monohydrochloride monohydrate, l-histidine, monohydrochloride, monohydrate, l-histidin hydrate hydrochloride, histidine, monohydrochloride, monohydrate, l, h-his-ohhclh2o, l-histidine, hydrochloride, monohydrate
PubChem CID 165377
IUPAC Name (2S)-2-amino-3-(1H-imidazol-4-yl)propanoic acid hydrate hydrochloride
SMILES O.Cl.N[C@@H](CC1=CNC=N1)C(O)=O

Specifications

Ammonium 200ppm max.
pH 3 to 5 (2.5% soln. at 25°C)
Color White
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Iron (Fe) 10ppm max.
Assay Percent Range 98%
Loss on Drying 7% to 10% (3 hrs) (145 to 150°C)
Other Amino Acids 0.5% max. (TLC)
Packaging Plastic Bottle
Beilstein 25, 513
Specific Rotation + 9.90
Sulfate 300ppm max.
Merck Index 10, 4617
Sulfated Ash 0.1% max.
Specific Rotation Condition + 9.90 (20.00°C c=11,6 N HCl)
Quantity 25 g
Formula Weight 209.64
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material L-Histidine hydrochloride monohydrate, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.