missing translation for 'onlineSavingsMsg'
Learn More

L-Histidine Hydrochloride Monohydrate (White Crystals or Crystalline Powder), Fisher BioReagents

Product Code. 11887693 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11887693 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11887693
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
L-Histidine Hydrochloride is suitable for use in tissue culture systems or as an amino acid standard.
TRUSTED_SUSTAINABILITY

Specifications

Ammonium 0.02% max.
CAS 5934-29-2
Arsenic (As) 1ppm max.
pH 3.0 to 5.0
Color White
Chloride 18.22 to 18.68
Heavy Metals (as Pb) 10ppm max.
Identification Pass Test
Loss on Drying 0.2% max.
Other Amino Acids Not Detected
Molecular Formula C6H12ClN3O3
Packaging Glass Bottle
MDL Number MFCD00151027
Residue after Ignition 0.1% max.
Specific Rotation 8.9 to 9.5° (+ or -)
State of Solution (%T) 98.0% min.
Synonym l-histidine hydrochloride hydrate, l-histidine hydrochloride monohydrate, s-2-amino-3-1h-imidazol-4-yl propanoic acid hydrochloride hydrate, h-his-oh.hcl.h2o, l-histidine monohydrochloride monohydrate, l-histidine, monohydrochloride, monohydrate, l-histidin hydrate hydrochloride, histidine, monohydrochloride, monohydrate, l, h-his-ohhclh2o, l-histidine, hydrochloride, monohydrate
Alpha Vector Histidine Hydrochloride Monohydrate
InChI Key CMXXUDSWGMGYLZ-XRIGFGBMSA-N
SMILES O.Cl.N[C@@H](CC1=CNC=N1)C(O)=O
IUPAC Name (2S)-2-amino-3-(1H-imidazol-4-yl)propanoic acid hydrate hydrochloride
Molecular Weight (g/mol) 209.63
Quantity 100 g
PubChem CID 165377
Formula Weight 209.64
Percent Purity 99.0 to 101.0%
Physical Form Solid
Chemical Name or Material L-Histidine Hydrochloride Monohydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.