missing translation for 'onlineSavingsMsg'
Learn More

L-Histidine, MedChemExpress

Product Code. 16422559
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16422559 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16422559 Supplier Medchem Express Supplier No. HYN0832
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress L-Histidine is an essential amino acid for infants. L-Histidine is an inhibitor of mitochondrial glutamine transport.

L-Histidine is an essential amino acid for infants. L-Histidine is an inhibitor of mitochondrial glutamine transport.

Specifications

Chemical Name or Material L-Histidine
CAS 71-00-1
Color White
Purity Grade Notes Research
Molecular Formula C6H9N3O2
Solubility Information H2O : 20.83 mg/mL (134.26 mM; ultrasonic and warming and heat to 80°C) ∣DMSO : 1 mg/mL (6.45 mM; ultrasonic and warming and heat to 80°C)
SMILES N[C@@H](CC1=CNC=N1)C(O)=O
Molecular Weight (g/mol) 155.15
Quantity 1 g
Formula Weight 155.15
Percent Purity 99.16%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.