missing translation for 'onlineSavingsMsg'
Learn More

L-Histidine Monohydrochloride Monohydrate, MP Biomedical

Product Code. 11354429 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
Unit Size:
100g
1kg
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11354429 1 kg 1kg
11324429 25 g 25g
11334429 100 g 100g
11344429 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11354429 Supplier MP Biomedicals Supplier No. 0210195791
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 5934-29-2
Molecular Formula C6H12ClN3O3
Molecular Weight (g/mol) 209.63
MDL Number MFCD00151027
InChI Key CMXXUDSWGMGYLZ-XRIGFGBMSA-N
Synonym l-histidine hydrochloride hydrate, l-histidine hydrochloride monohydrate, s-2-amino-3-1h-imidazol-4-yl propanoic acid hydrochloride hydrate, h-his-oh.hcl.h2o, l-histidine monohydrochloride monohydrate, l-histidine, monohydrochloride, monohydrate, l-histidin hydrate hydrochloride, histidine, monohydrochloride, monohydrate, l, h-his-ohhclh2o, l-histidine, hydrochloride, monohydrate
PubChem CID 165377
IUPAC Name (2S)-2-amino-3-(1H-imidazol-4-yl)propanoic acid hydrate hydrochloride
SMILES O.Cl.N[C@@H](CC1=CNC=N1)C(O)=O

Specifications

Color White
Quantity 1 kg
Formula Weight 209.6
Percent Purity 99.5%
Physical Form Powder
Chemical Name or Material L-Histidine Monohydrochloride Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.