missing translation for 'onlineSavingsMsg'
Learn More

L-Histidine Monohydrochloride, Multi-Compendial, F.C.C., J.T. Baker™

Product Code. 13284004 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
13284004 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13284004 Supplier Avantor J.T.Baker Supplier No. 208106
Quantity Price
1 465.00 € / 1kg
6 412.50 € / 1kg

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 5934-29-2
Molecular Formula C6H12ClN3O3
Packaging Poly Bottle
MDL Number MFCD00151027
Synonym l-histidine hydrochloride hydrate, l-histidine hydrochloride monohydrate, s-2-amino-3-1h-imidazol-4-yl propanoic acid hydrochloride hydrate, h-his-oh.hcl.h2o, l-histidine monohydrochloride monohydrate, l-histidine, monohydrochloride, monohydrate, l-histidin hydrate hydrochloride, histidine, monohydrochloride, monohydrate, l, h-his-ohhclh2o, l-histidine, hydrochloride, monohydrate
InChI Key CMXXUDSWGMGYLZ-XRIGFGBMSA-N
SMILES O.Cl.N[C@@H](CC1=CNC=N1)C(O)=O
IUPAC Name (2S)-2-amino-3-(1H-imidazol-4-yl)propanoic acid hydrate hydrochloride
Molecular Weight (g/mol) 209.63
Quantity 1 kg
PubChem CID 165377
Formula Weight 209.63
Grade FCC/Multi-Compendial
Chemical Name or Material L-Histidine Monohydrochloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.