missing translation for 'onlineSavingsMsg'
Learn More

L-Histidine, Pure, Fisher Chemical™

Product Code. 12696717 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
3 kg
Unit Size:
100g
3kg
Product Code. Quantity unitSize
12696717 3 kg 3kg
10161760 100 g 100g
2 options
This item is not returnable. View return policy

Product Code. 12696717

Brand: Fisher Chemical H/0467/63

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 71-00-1
Molecular Formula C6H9N3O2
Molecular Weight (g/mol) 155.16
MDL Number MFCD00064315
InChI Key HNDVDQJCIGZPNO-UHFFFAOYNA-N
Synonym l-histidine, histidine, h-his-oh, glyoxaline-5-alanine, anti-rheuma, l---histidine, istidina, s-histidine, l-histidin
PubChem CID 6274
ChEBI CHEBI:15971
IUPAC Name (2S)-2-amino-3-(1H-imidazol-5-yl)propanoic acid
SMILES NC(CC1=CN=CN1)C(O)=O

Specifications

Melting Point 282°C
pH 7.0 to 8.5
Color White
Packaging Plastic Powder Jar
Quantity 3 kg
Formula Weight 155.16g/mol
Physical Form Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.