missing translation for 'onlineSavingsMsg'
Learn More

L-Histidine, Pure, Fisher Chemical™

Product Code. p-7073065 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
3 kg
Unit Size:
100g
3kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10161760 100 g 100g
12696717 3 kg 3kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10161760 Supplier Fisher Chemical Supplier No. H/0467/48
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 71-00-1
Molecular Formula C6H9N3O2
Molecular Weight (g/mol) 155.16
MDL Number MFCD00064315
InChI Key HNDVDQJCIGZPNO-UHFFFAOYNA-N
Synonym l-histidine, histidine, h-his-oh, glyoxaline-5-alanine, anti-rheuma, l---histidine, istidina, s-histidine, l-histidin
PubChem CID 6274
ChEBI CHEBI:15971
IUPAC Name (2S)-2-amino-3-(1H-imidazol-5-yl)propanoic acid
SMILES NC(CC1=CN=CN1)C(O)=O

Specifications

Melting Point 282°C
pH 7.0 to 8.5
Color White
Packaging Plastic Powder Jar
Quantity 100 g
Formula Weight 155.16g/mol
Physical Form Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.