missing translation for 'onlineSavingsMsg'
Learn More

L-Histidine, TRC

Product Code. 30136499
Change view
Click to view available options
Quantity:
10 mg
5 g
50 g
Unit Size:
10mg
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136499 10 mg 10mg
30136001 50 g 50g
17969534 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136499 Supplier Toronto Research Chemicals Supplier No. TRCH45601010MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-00-1
Molecular Formula C6 H9 N3 O2
Molecular Weight (g/mol) 155.15
Synonym Histidine, (S)-2-Amino-3-(imidazol-4-yl)propanoic acid, L-Histidine (ACI), Histidine, L- (8CI), (2S)-2-Amino-3-(1H-imidazol-4-yl)propanoic acid, (2S)-2-Azaniumyl-3-(1H-imidazol-5-yl)propanoate, (S)-2-Amino-3-(1H-imidazol-4-yl)propanoic acid, (S)-4-(2-Amino-2-carboxyethyl)imidazole, (S)-Histidine, (S)-α-Amino-1H-imidazole-4-propanoic acid, 1H-Imidazole-4-alanine, (S)-, 1H-Imidazole-4-propanoic acid, α-amino-, (S)-, L-(-)-Histidine, L-Alanine, 3-(1H-imidazol-4-yl)-, Glyoxaline-5-alanine, NSC 137773
IUPAC Name (2S)-2-amino-3-(1H-imidazol-4-yl)propanoic acid
SMILES N[C@@H](Cc1c[nH]cn1)C(=O)O

Specifications

InChI Formula InChI=1S/C6H9N3O2/c7-5(6(10)11)1-4-2-8-3-9-4/h2-3,5H,1,7H2,(H,8,9)(H,10,11)/t5-/m0/s1
Quantity 10 mg
Formula Weight 155.0695
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material L-Histidine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.