missing translation for 'onlineSavingsMsg'
Learn More

L-Homoserine, 97%, Thermo Scientific™

Product Code. 10144640 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10144640 1 g Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10144640 Supplier Thermo Scientific Acros Supplier No. 204770010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 672-15-1
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
MDL Number MFCD00063090
InChI Key UKAUYVFTDYCKQA-VKHMYHEASA-N
Synonym l-homoserine, homoserine, s-2-amino-4-hydroxybutanoic acid, 2s-2-amino-4-hydroxybutanoic acid, h-hoser-oh, s-2-amino-4-hydroxybutyric acid, homoserine van, 2-amino-4-hydroxybutyric acid, s-homoserine, unii-6ka95x0ivo
PubChem CID 12647
ChEBI CHEBI:15699
IUPAC Name (2S)-2-amino-4-hydroxybutanoic acid
SMILES C(CO)C(C(=O)O)N

Specifications

Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Beilstein 04, III, 1636
Specific Rotation -8° to -9° (22°C, 589nm) (c=2, H2O)
Merck Index 15, 4778
Specific Rotation Condition − 8.50 (22.00°C c=2,H2O)
Quantity 1 g
Formula Weight 119.12
Percent Purity ≥96.0%
Physical Form Crystalline Powder
Chemical Name or Material L-Homoserine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.