missing translation for 'onlineSavingsMsg'
Learn More

L-Homoserine, 99%

Product Code. 11318805
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11318805 1 g 1g
11338805 250 mg 250mg
11328805 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11318805 Supplier Thermo Scientific Alfa Aesar Supplier No. L12427.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 672-15-1 | C4H9NO3 | 119.12 g/mol

L-Homoserine is used in the biosynthesis of methionine, threonine and isoleucine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Homoserine is used in the biosynthesis of methionine, threonine and isoleucine.

Solubility
Soluble in water (1100 mg/ml at 30°C), and methanol (Sparingly). Insoluble in ethanol.

Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 672-15-1
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
MDL Number MFCD00063090
InChI Key UKAUYVFTDYCKQA-VKHMYHEASA-N
Synonym l-homoserine, homoserine, s-2-amino-4-hydroxybutanoic acid, 2s-2-amino-4-hydroxybutanoic acid, h-hoser-oh, s-2-amino-4-hydroxybutyric acid, homoserine van, 2-amino-4-hydroxybutyric acid, s-homoserine, unii-6ka95x0ivo
PubChem CID 12647
ChEBI CHEBI:15699
IUPAC Name (2S)-2-amino-4-hydroxybutanoic acid
SMILES C(CO)C(C(=O)O)N

Specifications

Melting Point ∽203°C (decomposition)
Beilstein 1721681
Merck Index 14,4739
Solubility Information Soluble in water (1100mg/ml at 30°C),and methanol (Sparingly). Insoluble in ethanol.
Quantity 1 g
Formula Weight 119.12
Optical Rotation −9° (c=2 in Water)
Percent Purity 99%
Chemical Name or Material L-Homoserine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.