missing translation for 'onlineSavingsMsg'
Learn More

L-Hydroxyproline, for Biochemistry, Fisher Chemical

Product Code. 10356280 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Plastic powder jar
Unit Size:
5g
Product Code. Quantity Packaging unitSize
10356280 5 g Plastic powder jar 5g
1 options
This item is not returnable. View return policy

Product Code. 10356280

Brand: Fisher Chemical H/2415/44

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 51-35-4
Melting Point 274°C
Color White
pH 6
Packaging Plastic powder jar
Molecular Formula C5H9NO3
MDL Number 64320
Quantity 5 g
Synonym trans-4-hydroxy-l-proline, hydroxyproline, l-hydroxyproline, 2s,4r-4-hydroxypyrrolidine-2-carboxylic acid, 4-hydroxy-l-proline, l-4-hydroxyproline, hydroxy-l-proline, trans-4-hydroxyproline, h-hyp-oh, hypro
InChI Key PMMYEEVYMWASQN-DMTCNVIQSA-N
SMILES C1C(CNC1C(=O)O)O
IUPAC Name (2S,4R)-4-hydroxypyrrolidine-2-carboxylic acid
Molecular Weight (g/mol) 131.131
PubChem CID 5810
ChEBI CHEBI:18095
Formula Weight 131.13g/mol
Physical Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.