missing translation for 'onlineSavingsMsg'
Learn More

L-Isoleucine methyl ester hydrochloride, 97%, Thermo Scientific™

Product Code. 10832611 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10832611 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10832611 Supplier Thermo Scientific Acros Supplier No. 434910010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 18598-74-8
Melting Point 96°C to 100°C
Color Orange to White
Elemental Analysis 45.1 to 47.4% C, 7.5 to 7.9% N
Infrared Spectrum Authentic
Assay Percent Range 96.0 to max. 104% min. (Argentometry)
Packaging Glass Bottle
Molecular Formula C7H16ClNO2
MDL Number MFCD00038911
Quantity 1 g
Specific Rotation 27.5
Synonym l-isoleucine methyl ester hydrochloride, h-ile-ome.hcl, methyl l-isoleucinate hydrochloride, methyl l-isoleucinate hcl, methyl 2s,3s-2-amino-3-methylpentanoate hydrochloride, 2s,3s-methyl 2-amino-3-methylpentanoate hydrochloride, l-isoleucine, methyl ester, hydrochloride, l-isoleucine methyl ester hydrochloride;h-ile-ome.hcl, h-lle-ome.hcl, h-lle-ome?cl
InChI Key GGTBEWGOPAFTTH-GEMLJDPKSA-N
SMILES Cl.CC[C@H](C)[C@H](N)C(=O)OC
IUPAC Name methyl (2S,3S)-2-amino-3-methylpentanoate hydrochloride
Molecular Weight (g/mol) 181.66
PubChem CID 11171351
Formula Weight 181.66
Percent Purity 97%
Physical Form Crystalline Powder or Powder
Chemical Name or Material L-Isoleucine methyl ester hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.