missing translation for 'onlineSavingsMsg'
Learn More

L-Isoleucine Methyl Ester Hydrochloride, TRC

Product Code. 30407006
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30407006 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30407006 Supplier Toronto Research Chemicals Supplier No. TRCI82018025G
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-Isoleucine Methyl Ester Hydrochloride
CAS 18598-74-8
Molecular Formula C7H15NO2 . HCl
Quantity 25 g
Synonym Isoleucine Methyl Ester Hydrochloride, Methyl L-Isoleucinate Hydrochloride, Methyl Isoleucinate Hydrochloride
InChI Formula InChI=1S/C7H15NO2.ClH/c1-4-5(2)6(8)7(9)10-3;/h5-6H,4,8H2,1-3H3;1H/t5-,6-;/m0./s1
SMILES Cl.CC[C@H](C)[C@H](N)C(=O)OC
Molecular Weight (g/mol) 181.66
Formula Weight 181.087
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.