missing translation for 'onlineSavingsMsg'
Learn More

L-(+)-Lactic acid, >98%, MP Biomedicals™

Product Code. 11491681 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11491681 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11491681 Supplier MP Biomedicals Supplier No. 0219022810
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 79-33-4
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.078
InChI Key JVTAAEKCZFNVCJ-REOHCLBHSA-N
Synonym l-lactic acid, l-+-lactic acid, s-2-hydroxypropanoic acid, s-lactic acid, sarcolactic acid, s-2-hydroxypropionic acid, 2s-2-hydroxypropanoic acid, +-lactic acid, espiritin, tisulac
PubChem CID 107689
ChEBI CHEBI:422
IUPAC Name (2S)-2-hydroxypropanoic acid
SMILES CC(C(=O)O)O

Specifications

Color White
Density 1190 to 1250Kg/m3 at 20°C
Quantity 10 g
Physical Form Crystalline Powder To Liquid
Formula Weight 90.08
Chemical Name or Material L-(+)-Lactic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.