missing translation for 'onlineSavingsMsg'
Learn More

L-Lactic acid, anhydrous, 98%

Product Code. 11377986
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11377986 100 g 100g
11357986 5 g 5g
11367986 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11377986 Supplier Thermo Scientific Alfa Aesar Supplier No. L13242.22
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 79-33-4 | C3H6O3 | 90.078 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Useful chiral synthon, building block for depsipeptides.L-Lactic acid is an intermediate in the fermentation of sugar. It plays a vital role in several biochemical processes. It is a major ingredient in processed foods such as yogurt, cottage cheeses, koumiss and kombucha. It gives the sour flavor to sourdough breads. It finds application in pharmaceuticals to prepare water-soluble lactates, topical preparations and cosmetics. It acts as a food preservative, flavoring agent, curing agent, descaler, soap-scum remover and an anti-bacterial agent. It is an active component of lactated Ringer′s solution and Hartmann′s solution. Further, it is used for fluid resuscitation after blood loss due to trauma and surgery.

Solubility
Soluble in water.

Notes
Hygroscopic. Store in a cool place. Incompatible with strong bases, strong oxidizing agents, strong reducing agents and metals.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-33-4
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.078
MDL Number MFCD00064266
InChI Key JVTAAEKCZFNVCJ-REOHCLBHSA-N
Synonym l-lactic acid, l-+-lactic acid, s-2-hydroxypropanoic acid, s-lactic acid, sarcolactic acid, s-2-hydroxypropionic acid, 2s-2-hydroxypropanoic acid, +-lactic acid, espiritin, tisulac
PubChem CID 107689
ChEBI CHEBI:422
IUPAC Name (2S)-2-hydroxypropanoic acid
SMILES CC(C(=O)O)O

Specifications

Melting Point 52°C to 54°C
Density 1.22
pH 2.8
Boiling Point 119°C (12 mmHg)
Flash Point >110°C (230°F)
Odor Odorless
Quantity 100 g
Beilstein 1720251
Sensitivity Hygroscopic
Merck Index 14,5337
Solubility Information Soluble in water.
Formula Weight 90.08
Percent Purity 98%
Chemical Name or Material L-Lactic acid, anhydrous
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.