missing translation for 'onlineSavingsMsg'
Learn More

L-(+)-Lactic acid, ca 85-90% aq. soln.

Product Code. 11473497
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11473497 2500 g 2500g
11453497 100 g 100g
11463497 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11473497 Supplier Thermo Scientific Alfa Aesar Supplier No. A17268.0E
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 79-33-4 | C3H6O3 | 90.078 g/mol

It is most commonly used for fluid resuscitation after blood loss due to trauma, surgery, or burn injury. It is used in the agricultural, chemical, leather processing, pharmaceuticals, and cosmetics industries, used as an electroplating agent, food/feed additive, pH regulator, cleaning/washing agent, and tanning agent; used to flavor animal feeds; used as a solvent and intermediate in chemical production, as a pH regulator in fabric finishing, and in paints, coatings, soaps, and cleaning products; used to make large scale and fine chemicals, pulp-paper-paper products, food products, and plastic products, in mining, health services, agriculture-forestry-fishing, and building and construction work.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is most commonly used for fluid resuscitation after blood loss due to trauma, surgery, or burn injury. It is used in the agricultural, chemical, leather processing, pharmaceuticals, and cosmetics industries, used as an electroplating agent, food/feed additive, pH regulator, cleaning/washing agent, disinfectant, and tanning agent; used to flavor animal feeds; used as a solvent and intermediate in chemical production, as a pH regulator in fabric finishing, and in paints, coatings, soaps, and cleaning products; used to make large scale and fine chemicals, pulp-paper-paper products, food products, and plastic products, in mining, health services, agriculture-forestry-fishing, and building and construction work.

Solubility
Fully miscible in water. Soluble in water.

Notes
Store away from strong bases and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-33-4
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.078
MDL Number MFCD00064266
InChI Key JVTAAEKCZFNVCJ-REOHCLBHSA-N
Synonym l-lactic acid, l-+-lactic acid, s-2-hydroxypropanoic acid, s-lactic acid, sarcolactic acid, s-2-hydroxypropionic acid, 2s-2-hydroxypropanoic acid, #NAME?, espiritin, tisulac
PubChem CID 107689
ChEBI CHEBI:422
IUPAC Name (2S)-2-hydroxypropanoic acid
SMILES CC(C(=O)O)O

Specifications

pH 2.8
Melting Point 18°C
Density 1.209
Quantity 2500 g
Flash Point >110°C (230°F)
Odor Odorless
Optical Rotation -13° (c=2.5 in 1N NaOH)
Refractive Index 1.427
Beilstein 1720251
Merck Index 145337
Solubility Information Fully miscible in water. Soluble in water.
Formula Weight 90.08
Concentration or Composition (by Analyte or Components) ≈85 to 90% aq. Soln.
Chemical Name or Material L-(+)-Lactic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.