missing translation for 'onlineSavingsMsg'
Learn More

L-Lactic acid, sodium salt, 60 wt% solution in water

Product Code. 10124630 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10124630 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10124630 Supplier Thermo Scientific Acros Supplier No. 250300010
  • ONLINE EXCLUSIVE PRICE
    103.00 €
    100.00 € / 1 L
    Save 3.00 €
    2% Off
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 867-56-1 | C3H5NaO3 | 112.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-Lactic Acid, Sodium Salt
Name Note 60 wt% Solution in Water
CAS 867-56-1, 64-17-5
Density 1.3300g/mL
pH 6.5 to 8.5
Packaging Glass bottle
Molecular Formula C3H5NaO3
Linear Formula CH3CH(OH)CO2Na
Refractive Index 1.4220 to 1.4250
MDL Number MFCD00066576
Quantity 1 L
Merck Index 14,8635
Specific Gravity 1.33
Synonym methyl red hydrochloride, unii-rfa0w63690, 4-dimethylaminoazobenzene-2'-carboxylic acid hydrochloride, eaetho masculinei 2, methyl red, acs, 2-4-dimethylamino phenyl azo benzoic acid monohydrochloride, c15h15n3o2.hcl, 2-e-4-dimethylamino phenyl diazenyl benzoic acid hydrochloride 1:1, methyl red hydrochloride, acs reagent, 2-4-dimethylaminophenyl diazenyl benzoic acid hydrochloride
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol
InChI Key ICTWKXAVJAHRMK-UHFFFAOYSA-N
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)O.Cl
IUPAC Name 2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid;hydrochloride
Molecular Weight (g/mol) 112.06
PubChem CID 113227
Viscosity 80-160 mPa.s (20°C)
Formula Weight 112.06
Percent Purity 59 to 61% (Total base)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.