missing translation for 'onlineSavingsMsg'
Learn More

L-Lactic acid sodium salt, 99%, extra pure

Product Code. 11985051 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11985051 10 g Glass bottle 10g
11995051 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11985051 Supplier Thermo Scientific Acros Supplier No. 439220100
From Chemicals to Chromatography
Use promo code 27991
View all promotions
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 867-56-1 | C3H5NaO3 | 112.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 867-56-1
Molecular Formula C3H5NaO3
Molecular Weight (g/mol) 112.06
Synonym sodium l-lactate, sodium-l-lactate, sodium lactate, l, sodium s-2-hydroxypropanoate, unii-p2y1c6m9ps, sodium s-lactate, sodium l-+-lactate, p2y1c6m9ps, propanoic acid, 2-hydroxy-, monosodium salt, 2s, l-+-lactic acid sodium salt

Specifications

Melting Point 161.6°C to 165°C
Color White to Yellow
Physical Form Powder
Quantity 10 g
Assay Percent Range 99%
Linear Formula CH3CH(OH)CO2Na
Formula Weight 112.06
Percent Purity 99%
Grade Extra Pure
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material L-Lactic acid sodium salt, Extra Pure
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.