missing translation for 'onlineSavingsMsg'
Learn More

L-Lactide, 98+%

Product Code. 11312846
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11312846 5 g 5g
11322846 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11312846 Supplier Thermo Scientific Alfa Aesar Supplier No. L09031.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4511-42-6 | C6H8O4 | 144.13 g/mol

L-Lactide upon polymerization gives poly(lactic acid), which finds many useful properties like biodegradable and nontoxic to the human body and the environment. Its polymer, poly-l-lactide is utilized in the repairing of orbital posterior floor fracture. Further, it is used as biomedical materials for tissue regeneration, matrices for drug delivery systems and alternatives for commercial polymeric materials to reduce the impact on the environment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in chloroform, and methanol. Slightly soluble in benzene.

Notes
Moisture sensitive. Store in a cool place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4511-42-6
Molecular Formula C6H8O4
Molecular Weight (g/mol) 144.13
MDL Number MFCD00070594
InChI Key JJTUDXZGHPGLLC-UHFFFAOYNA-N
Synonym l-lactide, 3s,6s-3,6-dimethyl-1,4-dioxane-2,5-dione, l---lactide, lactide, l, l-dilactide, 3s-cis-3,6-dimethyl-1,4-dioxane-2,5-dione, ll-lactide, unii-ij13to4no1, 1,4-dioxane-2,5-dione, 3,6-dimethyl-, 3s,6s, ij13to4no1
PubChem CID 107983
SMILES CC1OC(=O)C(C)OC1=O

Specifications

pH 3.0
Melting Point 95°C to 99°C
Boiling Point 255°C
Quantity 5 g
Refractive Index 1.4475
Beilstein 82124
Solubility Information Soluble in chloroform,and methanol. Slightly soluble in benzene.
IUPAC Name 3,6-dimethyl-1,4-dioxane-2,5-dione
Formula Weight 144.13
Percent Purity ≥98%
Chemical Name or Material L-Lactide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.